2-Fluor-5-methyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-methylpyridine-3-boronic acid, (2-Fluoro-5-methylpyridin-3-yl)boronic acid

BB-8519-1G
COBBBB-8519-1GEA 497 EUR
COBBBB-8519-1G
2-Fluor-5-methyl-3-pyridinboronsäure
2-Fluoro-5-methyl-3-pyridineboronic acid
Formel: C₆H₇BFNO₂
Molecular Weight: 154,940 g/mol
MDL: MFCD07368857
CAS-Nummer: 1072952-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR