2-Methoxy-4-methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methoxy-4-methylpyridin-3-ylboronic acid, 2-Methoxy-4-methylpyridine-5-boronic acid

BB-8521-1G
COBBBB-8521-1GEA 249 EUR
COBBBB-8521-1G
2-Methoxy-4-methyl-5-pyridinboronsäure
2-Methoxy-4-methyl-5-pyridinboronsäure
Formel: C₇H₁₀BNO₃
Molecular Weight: 166,97 g/mol
MDL: MFCD07368880
CAS-Nummer: 503184-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR