2-Amino-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminopyrimidine-5-boronic acid, (2-Aminopyrimidin-5-yl)boronic acid

BB-8524-1G
COBBBB-8524-1GEA 50.9 EUR
COBBBB-8524-1G
2-Amino-5-pyrimidinboronsäure
2-Amino-5-pyrimidineboronic acid
Formel: C₄H₆BN₃O₂
Molecular Weight: 138,92 g/mol
MDL: MFCD07375147
CAS-Nummer: 936250-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR