2-(Trifluormethyl)-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid
BB-8525-5G
BB-8525-1G
COBBBB-8525-5GEA
249
EUR
COBBBB-8525-5G
COBBBB-8525-1G
2-(Trifluormethyl)-5-pyridinboronsäure
2-(Trifluormethyl)-5-pyridinboronsäure
Formel:
C₆H₅BF₃NO₂ Molecular Weight: 190.92 g/mol |
MDL:
MFCD07375381 CAS-Nummer: 868662-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...