2-(Trifluormethyl)-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid

BB-8525-5G BB-8525-1G
COBBBB-8525-5GEA 249 EUR
COBBBB-8525-5G COBBBB-8525-1G
2-(Trifluormethyl)-5-pyridinboronsäure
2-(Trifluormethyl)-5-pyridinboronsäure
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190.92 g/mol
MDL: MFCD07375381
CAS-Nummer: 868662-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR