4-Amino-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8526-1G
COBBBB-8526-1GEA 0 EUR
COBBBB-8526-1G
4-Amino-3-nitrophenylboronsäure
4-Amino-3-nitrophenylboronsäure
Formel: C₆H₇BN₂O₄
MDL: MFCD07437851
CAS-Nummer: 89466-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR