3-Chlor-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-5-(trifluoromethyl)phenyl)boronic acid

BB-8529-5G BB-8529-1G
COBBBB-8529-5GEA 169 EUR
COBBBB-8529-5G COBBBB-8529-1G
3-Chlor-5-(trifluormethyl)phenylboronsäure
3-Chlor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
Molecular Weight: 224.3700 g/mol
MDL: MFCD07778020
CAS-Nummer: 1160561-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR