3-(Ethoxycarbonyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Ethoxycarbonyl-2-fluorobenzeneboronic acid, 2-Fluoro-3-(ethoxycarbonyl)phenylboronic acid, (3-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid

BB-8543-5G BB-8543-1G
COBBBB-8543-5GEA 0 EUR
COBBBB-8543-5G COBBBB-8543-1G
3-(Ethoxycarbonyl)-2-fluorphenylboronsäure
3-(Ethoxycarbonyl)-2-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
MDL: MFCD08064050
CAS-Nummer: 1072952-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR