3-Chlor-4-formylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-4-formylbenzeneboronic acid, (3-Chloro-4-formylphenyl)boronic acid
BB-8545-1G
COBBBB-8545-1GEA
298
EUR
COBBBB-8545-1G
3-Chlor-4-formylphenylboronsäure
3-Chlor-4-formylphenylboronsäure
Formel:
C₇H₆BClO₃ Molecular Weight: 184,39 g/mol |
MDL:
MFCD08274473 CAS-Nummer: 1072952-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...