2,3-Dichlor-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,3-Dichloropyridine-5-boronic acid

BB-8550-1G
COBBBB-8550-1GEA 262 EUR
COBBBB-8550-1G
2,3-Dichlor-5-pyridinboronsäure
2,3-Dichlor-5-pyridinboronsäure
Formel: C₅H₄BCl₂NO₂
Molecular Weight: 191,81 g/mol
MDL: MFCD08277308
CAS-Nummer: 1072944-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR