3-Brom-2-chlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-chlorpyridin-4-boronsäure, 3-Brom-2-chlor-4-pyridinylboronsäure

BB-8551-1G
COBBBB-8551-1GEA 153 EUR
COBBBB-8551-1G
3-Brom-2-chlor-4-pyridinboronsäure
3-Bromo-2-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
MDL: MFCD08458470
CAS-Nummer: 1072944-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR