2-Fluor-4-methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-methylpyridine-5-boronic acid

BB-8568-5G BB-8568-1G
COBBBB-8568-5GEA 193 EUR
COBBBB-8568-5G COBBBB-8568-1G
2-Fluor-4-methyl-5-pyridinboronsäure
2-Fluor-4-methyl-5-pyridinboronsäure
Formel: C₆H₇BFNO₂
Molecular Weight: 154.9400 g/mol
MDL: MFCD08741406
CAS-Nummer: 1072944-18-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR