5-Brom-2-chlor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlorpyridin-3-boronsäure, 5-Brom-2-chlor-3-pyridinylboronsäure

BB-8571-250M
COBBBB-8571-250MEA 257 EUR
COBBBB-8571-250M
5-Brom-2-chlor-3-pyridinboronsäure
5-Bromo-2-chloro-3-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
MDL: MFCD09037486
CAS-Nummer: 1072944-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR