2-(Methylthio)-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8573-250M
COBBBB-8573-250MEA 303 EUR
COBBBB-8573-250M
2-(Methylthio)-3-pyridinboronsäure
2-(Methylthio)-3-pyridineboronic acid
Formel: C₆H₈BNO₂S
Molecular Weight: 169,01 g/mol
MDL: MFCD09037488
CAS-Nummer: 1072944-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR