1-(2-Tetrahydropyranyl)-1H-pyrazol-4-boronsäureneopentylglycolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

BB-8578-250M
COBBBB-8578-250MEA 114 EUR
COBBBB-8578-250M
1-(2-Tetrahydropyranyl)-1H-pyrazol-4-boronsäureneopentylglycolester
1-(2-Tetrahydropyranyl)-1H-pyrazol-4-boronsäureneopentylglycolester
Formel: C₁₃H₂₁BN₂O₃
Molecular Weight: 264,13 g/mol
MDL: MFCD09037503
CAS-Nummer: 1072944-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR