4-(2-Naphthyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Naphthalen-2-yl)phenyl)boronic acid

BB-8582-1G
COBBBB-8582-1GEA 65.8 EUR
COBBBB-8582-1G
4-(2-Naphthyl)phenylboronsäure
4-(2-Naphthyl)phenylboronsäure
Formel: C₁₆H₁₃BO₂
Molecular Weight: 248,0800 g/mol
MDL: MFCD09260454
CAS-Nummer: 918655-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR