1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäure

Lieferant: COMBI-BLOCKS

BB-8782-1G
COBBBB-8782-1GEA 0 EUR
COBBBB-8782-1G
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäure
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäure
Formel: C₉H₁₉BN₂O₃Si
MDL: MFCD11855854
CAS-Nummer: 188978-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR