1-Methyl-1H-pyrazol-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-Methyl-1H-pyrazol-5-yl)boronic acid, (1-Methyl-1H-pyrazol-5-yl)-boronic acid

BB-8865-1G
COBBBB-8865-1GEA 0 EUR
COBBBB-8865-1G
1-Methyl-1H-pyrazol-5-boronsäure
1-Methyl-1H-pyrazol-5-boronsäure
Formel: C₄H₇BN₂O₂
Molecular Weight: 125,92 g/mol
MDL: MFCD11053031
CAS-Nummer: 720702-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR