4-Isoxazolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Isoxazol-4-ylboronic acid

BB-8875-1G
COBBBB-8875-1GEA 485 EUR
COBBBB-8875-1G
4-Isoxazolboronsäure
Isoxazole-4-boronic acid
Formel: C₃H₄BNO₃
Molecular Weight: 112.8798 g/mol
MDL: MFCD06657892
CAS-Nummer: 1008139-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR