6-Brom-2-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Bromopyridin-2-yl)boronic acid

BB-8882-5G BB-8882-1G
COBBBB-8882-5GEA 1070 EUR
COBBBB-8882-5G COBBBB-8882-1G
6-Brom-2-pyridinboronsäure
6-Bromo-2-pyridineboronic acid
Formel: C₅H₅BBrNO₂
Molecular Weight: 201.81 g/mol
MDL: MFCD04114287
CAS-Nummer: 440680-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR