2-Morpholino-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-8925-1G
COBBBB-8925-1GEA 0 EUR
COBBBB-8925-1G
2-Morpholino-3-pyridinboronsäure
2-Morpholino-3-pyridinboronsäure
Formel: C₉H₁₃BN₂O₃
MDL: MFCD11867876
CAS-Nummer: 1218790-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR