4-(Ethoxycarbonylmethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-[(Ethoxycarbonyl)methyl]phenylboronic acid, (4-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid

BB-9006-1G BB-9006-250M
COBBBB-9006-1GEA 852 EUR
COBBBB-9006-1G COBBBB-9006-250M
4-(Ethoxycarbonylmethyl)phenylboronsäure
4-(Ethoxycarbonylmethyl)phenylboronsäure
Formel: C₁₀H₁₃BO₄
Molecular Weight: 208.200 g/mol
MDL: MFCD08704683
CAS-Nummer: 92243-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR