1-Methyl-1H-indazol-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-1H-indazol-5-yl-5-boronic acid, (1-methyl-1H-indazol-5-yl)boronic acid

BB-9020-1G
COBBBB-9020-1GEA 298 EUR
COBBBB-9020-1G
1-Methyl-1H-indazol-5-boronsäure
1-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9800 g/mol
MDL: MFCD09870052
CAS-Nummer: 590418-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR