1-Methyl-1H-indazol-6-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Methyl-1H-indazole-6-boronic Acid, 1-Methylindazole-6-boronic acid

BB-9024-5G
COBBBB-9024-5GEA 624 EUR
COBBBB-9024-5G
1-Methyl-1H-indazol-6-boronsäure
1-Methyl-1H-indazole-6-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9803 g/mol
MDL: MFCD09870053
CAS-Nummer: 1150114-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR