3-Fluor-4-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-9047-5G BB-9047-1G
COBBBB-9047-5GEA 0 EUR
COBBBB-9047-5G COBBBB-9047-1G
3-Fluor-4-(trifluormethoxy)phenylboronsäure
3-Fluor-4-(trifluormethoxy)phenylboronsäure
Formel: C₇H₅BF₄O₃
MDL: MFCD07368680
CAS-Nummer: 187804-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR