3,4-Difluor-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,4-Difluoro-5-methoxyphenyl)boronic acid

BB-9054-5G BB-9054-1G
COBBBB-9054-5GEA 0 EUR
COBBBB-9054-5G COBBBB-9054-1G
3,4-Difluor-5-methoxyphenylboronsäure
3,4-Difluor-5-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
MDL: MFCD09907960
CAS-Nummer: 925910-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR