4-Brom-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromonaphthalen-1-yl)boronic acid

BB-9079-1G
COBBBB-9079-1GEA 75.9 EUR
COBBBB-9079-1G
4-Brom-1-naphthalinboronsäure
4-Bromo-1-naphthaleneboronic acid
Formel: C₁₀H₈BBrO₂
Molecular Weight: 250,88 g/mol
MDL: MFCD08701764
CAS-Nummer: 145965-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR