1H-Indazol-4-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid
BB-9080-1G
COBBBB-9080-1GEA
154
EUR
COBBBB-9080-1G
1H-Indazol-4-boronsäure
1H-Indazole-4-boronic acid
Formel:
C₇H₇BN₂O₂ Molecular Weight: 161,9500 g/mol |
MDL:
MFCD09878901 CAS-Nummer: 1023595-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...