3,5-Dibromtoluol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
1,3-Dibromo-5-methylbenzene
BK-4509-25G
BK-4509-5G
BK-4509-100G
COBBBK-4509-25GEA
0
EUR
COBBBK-4509-25G
COBBBK-4509-5G
COBBBK-4509-100G
3,5-Dibromtoluol
3,5-Dibromtoluol
Formel:
C₇H₆Br₂ |
MDL:
MFCD00013528 CAS-Nummer: 1611-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...