3,5-Dibromtoluol

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 1,3-Dibromo-5-methylbenzene

BK-4509-25G BK-4509-5G BK-4509-100G
COBBBK-4509-25GEA 0 EUR
COBBBK-4509-25G COBBBK-4509-5G COBBBK-4509-100G
3,5-Dibromtoluol
3,5-Dibromtoluol
Formel: C₇H₆Br₂
MDL: MFCD00013528
CAS-Nummer: 1611-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR