2-Chlorisonicotinonitril

Lieferant: COMBI-BLOCKS

BK-4524-100G BK-4524-25G
COBBBK-4524-100GEA 0 EUR
COBBBK-4524-100G COBBBK-4524-25G
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00174318
CAS-Nummer: 33252-30-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR