2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat

Lieferant: COMBI-BLOCKS

BO-2238-1G
COBBBO-2238-1GEA 0 EUR
COBBBO-2238-1G
2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat
2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat
Formel: C₁₂H₂₁BN₂O₄·H₂O
MDL: MFCD02180675
CAS-Nummer: 306935-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR