2-Brom-5-fluorbenzoesäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
CA-4003-25G
CA-4003-100G
COBBCA-4003-25GEA
0
EUR
COBBCA-4003-25G
COBBCA-4003-100G
2-Brom-5-fluorbenzoesäure
2-Bromo-5-fluorobenzoic acid
Formel:
C₇H₄BrFO₂ |
MDL:
MFCD00142874 CAS-Nummer: 394-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...