2-Aminonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Aminopyridin-3-carbonsäure, 2-Amino-3-pyridincarbonsäure
CA-4056-100G
CA-4056-25G
COBBCA-4056-100GEA
0
EUR
COBBCA-4056-100G
COBBCA-4056-25G
2-Aminonicotinsäure
2-Aminonicotinsäure
Formel:
C₆H₆N₂O₂ Schmelzpunkt: 297 °C Dichte: 1,545 g/cm³ (20 °C) |
MDL:
MFCD00006318 CAS-Nummer: 5345-47-1 EINECS: 226-296-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...