5-Aminonicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid

CA-4060-5G CA-4060-25G CA-4060-1G
COBBCA-4060-5GEA 35.9 EUR
COBBCA-4060-5G COBBCA-4060-25G COBBCA-4060-1G
5-Aminonicotinsäure
5-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
MDL: MFCD00129116
CAS-Nummer: 24242-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR