4-Aminonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure
CA-4067-5G
CA-4067-25G
CA-4067-1G
COBBCA-4067-5GEA
50.9
EUR
COBBCA-4067-5G
COBBCA-4067-25G
COBBCA-4067-1G
4-Aminonicotinsäure
4-Aminonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138.13 g/mol |
MDL:
MFCD00234183 CAS-Nummer: 7418-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...