Methyl-6-aminonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

CA-4102-5G CA-4102-25G CA-4102-1G
COBBCA-4102-5GEA 51 EUR
COBBCA-4102-5G COBBCA-4102-25G COBBCA-4102-1G
Methyl-6-aminonicotinat
Methyl-6-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
MDL: MFCD00797844
CAS-Nummer: 36052-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR