3-Acetamido-5-brombenzoesäure

Lieferant: COMBI-BLOCKS

CA-4115-5G CA-4115-1G
COBBCA-4115-5GEA 0 EUR
COBBCA-4115-5G COBBCA-4115-1G
3-Acetamido-5-brombenzoesäure
3-Acetamido-5-brombenzoesäure
Formel: C₉H₈BrNO₃
MDL: MFCD01098204
CAS-Nummer: 78238-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR