Methyl-5-brompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

CA-4117-5G CA-4117-25G CA-4117-1G
COBBCA-4117-5GEA 97.4 EUR
COBBCA-4117-5G COBBCA-4117-25G COBBCA-4117-1G
Methyl-5-brompicolinat
Methyl-5-brompicolinat
Formel: C₇H₆BrNO₂
Molecular Weight: 216.03 g/mol
MDL: MFCD04112493
CAS-Nummer: 29682-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR