Methyl-6-hydroxy-5-nitronicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-hydroxy-5-nitronicotinate, Methyl 6-hydroxy-5-nitropyridine-3-carboxylate

CA-4121-5G CA-4121-1G
COBBCA-4121-5GEA 576 EUR
COBBCA-4121-5G COBBCA-4121-1G
Methyl-6-hydroxy-5-nitronicotinat
Methyl-6-hydroxy-5-nitronicotinat
Formel: C₇H₆N₂O₅
Molecular Weight: 198.14 g/mol
MDL: MFCD10699641
CAS-Nummer: 222970-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR