2-Hydroxynicotinsäure

Lieferant: COMBI-BLOCKS

CA-4124-25G CA-4124-100G
COBBCA-4124-25GEA 0 EUR
COBBCA-4124-25G COBBCA-4124-100G
2-Hydroxynicotinsäure
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR