2-Amino-5-bromisonicotinsäure

Lieferant: COMBI-BLOCKS

CA-4139-5G CA-4139-1G
COBBCA-4139-5GEA 191 EUR
COBBCA-4139-5G COBBCA-4139-1G
2-Amino-5-bromisonicotinsäure
2-Amino-5-bromisonicotinsäure
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.0200 g/mol
MDL: MFCD09878357
CAS-Nummer: 1000339-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR