Methyl-4-hydroxypicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-hydroxypicolinate

CA-4149-5G CA-4149-1G CA-4149-250M
COBBCA-4149-5GEA 1330 EUR
COBBCA-4149-5G COBBCA-4149-1G COBBCA-4149-250M
Methyl-4-hydroxypicolinat
Methyl-4-hydroxypicolinat
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
MDL: MFCD12025998
CAS-Nummer: 473269-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR