2-Chlor-5-nitronicotinsäure

Lieferant: COMBI-BLOCKS

CA-4174-5G CA-4174-1G
COBBCA-4174-5GEA 0 EUR
COBBCA-4174-5G COBBCA-4174-1G
2-Chlor-5-nitronicotinsäure
2-Chlor-5-nitronicotinsäure
Formel: C₆H₃ClN₂O₄
MDL: MFCD06658402
CAS-Nummer: 42959-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR