2-Chlor-5-nitronicotinsäure
Lieferant: COMBI-BLOCKS
CA-4174-5G
CA-4174-1G
COBBCA-4174-5GEA
0
EUR
COBBCA-4174-5G
COBBCA-4174-1G
2-Chlor-5-nitronicotinsäure
2-Chlor-5-nitronicotinsäure
Formel:
C₆H₃ClN₂O₄ |
MDL:
MFCD06658402 CAS-Nummer: 42959-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...