3-(Benzyloxy)-4-brombenzoesäure

Lieferant: COMBI-BLOCKS

CA-4193-5G
COBBCA-4193-5GEA 595 EUR
COBBCA-4193-5G
3-(Benzyloxy)-4-brombenzoesäure
3-(Benzyloxy)-4-brombenzoesäure
Formel: C₁₄H₁₁BrO₃
Molecular Weight: 307,139 g/mol
MDL: MFCD08436069
CAS-Nummer: 17054-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR