3-Hydroxy-4-nitrobenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

CA-4196-25G CA-4196-100G CA-4196-5G
COBBCA-4196-25GEA 47.5 EUR
COBBCA-4196-25G COBBCA-4196-100G COBBCA-4196-5G
3-Hydroxy-4-nitrobenzoesäure
3-Hydroxy-4-nitrobenzoic acid
Formel: C₇H₅NO₅
Molecular Weight: 183.12 g/mol
Schmelzpunkt: 234…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007110
CAS-Nummer: 619-14-7
EINECS: 210-580-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR