4-Fluor-3-hydroxybenzonitril

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluor-3-hydroxybenzoesäurenitril

CA-4233-5G CA-4233-1G
COBBCA-4233-5GEA 0 EUR
COBBCA-4233-5G COBBCA-4233-1G
4-Fluor-3-hydroxybenzonitril
4-Fluor-3-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137.11 g/mol
MDL: MFCD08689519
CAS-Nummer: 186590-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR