3-Brom-o-anisaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-2-methoxybenzaldehyde

CA-4234-5G CA-4234-1G
COBBCA-4234-5GEA 543 EUR
COBBCA-4234-5G COBBCA-4234-1G
3-Brom-o-anisaldehyd
3-Brom-o-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.0400 g/mol
MDL: MFCD09834811
CAS-Nummer: 88275-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR