6-Nitro-m-anissäure

Lieferant: COMBI-BLOCKS

CA-4267-5G CA-4267-25G CA-4267-100G
COBBCA-4267-5GEA 0 EUR
COBBCA-4267-5G COBBCA-4267-25G COBBCA-4267-100G
6-Nitro-m-anissäure
6-Nitro-m-anissäure
Formel: C₈H₇NO₅
Schmelzpunkt: 138 °C
MDL: MFCD00151836
CAS-Nummer: 1882-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR